siRNA Details
| Gene Symbol: RPS28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031.4 | NP_001022.1 | 1 | 370 | ||
| GenBank mRNA | AK293636.1 | - | 3 | 619 | ||
| AK301638.1 | - | 2 | 799 | |||
| AU126783.1 | - | 4 | 370 | |||
| BC000354.1 | - | 1 | 356 | 2964364 | ||
| BC018810.1 | - | 1 | 356 | |||
| BC021239.2 | - | 1 | 349 | 4309752 | ||
| BC070217.1 | - | 1 | 372 | 4248660 | ||
| BC070218.1 | - | 1 | 367 | 6649143 | ||
| L05091.1 | - | 4 | 354 | |||
| U58682.1 | - | 4 | 351 | |||

RPS28
Genomic Map