siRNA Details
| Gene Symbol: RPS28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031.4 | NP_001022.1 | 1 | 265 | ||
| GenBank mRNA | AK293636.1 | - | 2, 3 | 514 | ||
| AU126783.1 | - | 3, 4 | 265 | |||
| BC000354.1 | - | 1 | 251 | 2964364 | ||
| BC018810.1 | - | 1 | 251 | |||
| BC021239.2 | - | 1 | 244 | 4309752 | ||
| BC070217.1 | - | 1 | 265 | 4248660 | ||
| BC070218.1 | - | 1 | 262 | 6649143 | ||
| L05091.1 | - | 3, 4 | 249 | |||

RPS28
Genomic Map