siRNA Details
| Gene Symbol: CBY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164444.1 | NP_001157916.1 | 2 | 556 | ||
| XM_017009726.1 | XP_016865215.1 | 2 | 1409 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CBY3| Gene Symbol: CBY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164444.1 | NP_001157916.1 | 2 | 556 | ||
| XM_017009726.1 | XP_016865215.1 | 2 | 1409 | |||