siRNA Details
| Gene Symbol: LOC107985803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017005446.1 | XP_016860935.1 | 5 | 593 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107985803| Gene Symbol: LOC107985803 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017005446.1 | XP_016860935.1 | 5 | 593 | ||