siRNA Details
| Gene Symbol: LINC01587 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126517.1 | 2 | 419 | |||
| NR_126518.1 | 2 | 398 | ||||
| NR_126519.1 | 3 | 307 | ||||
| GenBank mRNA | BC117444.1 | - | 2 | 352 | 40125995 | |
| BC117446.1 | - | 2 | 352 | 40125997 | ||
| BX094519.1 | - | 2 | 216 | |||
| D82070.1 | - | 2 | 398 | |||
| DB043858.1 | - | 3 | 307 | |||

LINC01587
Genomic Map