siRNA Details
| Gene Symbol: GOLGA6L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC101294.1 | - | 2 | 267 | ||
| Gene Symbol: GOLGA6L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC101293.1 | - | 2 | 249 | ||

GOLGA6L4
Genomic Map