siRNA Details
| Gene Symbol: LOC101929322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011516748.2 | XP_011515050.1 | 2 | 491 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929322| Gene Symbol: LOC101929322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011516748.2 | XP_011515050.1 | 2 | 491 | ||