siRNA Details
| Gene Symbol: TEX22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195082.1 | NP_001182011.1 | 4 | 500 | ||
| XM_006720234.3 | XP_006720297.1 | 4 | 774 | |||
| GenBank mRNA | BX378619.2 | - | 4 | 500 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TEX22| Gene Symbol: TEX22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195082.1 | NP_001182011.1 | 4 | 500 | ||
| XM_006720234.3 | XP_006720297.1 | 4 | 774 | |||
| GenBank mRNA | BX378619.2 | - | 4 | 500 | ||