siRNA Details
| Gene Symbol: SNRPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| GenBank mRNA | AF400493.1 | - | LIT2215 IC-SNURF-SNRPN 3 prime noncoding transcripts |
2 | 72 | |
| AY362864.1 | - | 5 | 357 | |||

SNRPN
Genomic Map