siRNA Details
| Gene Symbol: NDOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144026.2 | NP_001137498.1 | 14 | 3647 | ||
| NM_001144027.2 | NP_001137499.1 | 13 | 3496 | |||
| NM_001144028.2 | NP_001137500.1 | 14 | 3599 | |||
| NM_014434.3 | NP_055249.1 | 14 | 3620 | |||
| XM_011518545.1 | XP_011516847.1 | 13 | 3535 | |||
| XM_011518547.1 | XP_011516849.1 | 14 | 3437 | |||
| XM_011518548.2 | XP_011516850.1 | 13 | 4837 | |||
| XM_017014627.1 | XP_016870116.1 | 14 | 3598 | |||
| GenBank mRNA | AL133632.1 | - | 1 | 72 | ||

NDOR1
Genomic Map