siRNA Details
| Gene Symbol: EBI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005755.2 | NP_005746.2 | 5 | 950 | ||
| XM_011527619.2 | XP_011525921.1 | 6 | 1199 | |||
| GenBank mRNA | BC015364.1 | - | 5 | 995 | 3897136 | |
| BC046112.1 | - | 5 | 949 | 5748376 | ||
| L08187.1 | - | 5 | 955 | |||

EBI3
Genomic Map