siRNA Details
| Gene Symbol: LOC100128108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321551.1 | NP_001308480.1 | 3 | 419 | ||
| NM_001321552.1 | NP_001308481.1 | 3 | 433 | |||
| NM_001321553.1 | NP_001308482.1 | 2 | 502 | |||
| XM_006720812.3 | XP_006720875.1 | 3 | 1423 | |||
| XM_017021852.1 | XP_016877341.1 | 3 | 760 | |||
| GenBank mRNA | BG206946.1 | - | 1 | 176 | ||

LOC100128108
Genomic Map