siRNA Details
Gene Symbol: ADGRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079858.2 | NP_001073327.1 | 22, 23 | 2089 | ||
NM_001079859.2 | NP_001073328.1 | 21, 22 | 2047 | |||
NM_001079860.2 | NP_001073329.1 | 20, 21 | 2023 | |||
NM_001184833.1 | NP_001171762.1 | 21, 22 | 2041 | |||
NM_001184834.1 | NP_001171763.1 | 22, 23 | 2089 | |||
NM_001184835.1 | NP_001171764.1 | 19, 20 | 1975 | |||
NM_001184836.1 | NP_001171765.1 | 20, 21 | 2017 | |||
NM_001184837.1 | NP_001171766.1 | 20, 21 | 1999 | |||
NM_005756.3 | NP_005747.2 | 22, 23 | 2080 | |||
XM_006724455.3 | XP_006724518.1 | 22, 23 | 1947 | |||
XM_011545434.1 | XP_011543736.1 | 22, 23 | 1912 | |||
XM_011545435.2 | XP_011543737.1 | 22, 23 | 2642 | |||
GenBank mRNA | AF538954.1 | - | 23, 24 | 2010 | ||
AF539455.1 | - | 21, 22 | 1920 | |||
AF539456.1 | - | 21, 22 | 1944 | |||
AK291012.1 | - | 22, 23 | 1924 | |||
AY143364.1 | - | 20, 21 | 1896 | |||
AY143365.1 | - | 22, 23 | 1968 | |||
AY143366.1 | - | 22, 23 | 1962 | |||
AY143367.1 | - | 21, 22 | 1938 | |||
AY148343.1 | - | 23, 24 | 2010 | |||
BC099901.1 | - | 21, 22 | 1874 | 30349668 | ||
BC113978.1 | - | 20, 21 | 1842 | 40081713 | ||
BC113979.1 | - | 17, 18 | 1533 | 40081711 | ||
X81892.1 | - | 23, 24 | 1910 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
