siRNA Details
| Gene Symbol: RPS28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031.4 | NP_001022.1 | 1 | 369 | ||
| GenBank mRNA | AK293636.1 | - | 3 | 618 | ||
| AK301638.1 | - | 2 | 798 | |||
| AU126783.1 | - | 4 | 369 | |||
| BC000354.1 | - | 1 | 355 | 2964364 | ||
| BC018810.1 | - | 1 | 355 | |||
| BC021239.2 | - | 1 | 348 | 4309752 | ||
| BC070217.1 | - | 1 | 371 | 4248660 | ||
| BC070218.1 | - | 1 | 366 | 6649143 | ||
| L05091.1 | - | 4 | 353 | |||
| U58682.1 | - | 4 | 350 | |||

RPS28
Genomic Map