siRNA Details
| Gene Symbol: RPS28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031.4 | NP_001022.1 | 1 | 300 | ||
| GenBank mRNA | AK301638.1 | - | 2 | 729 | ||
| BC021239.2 | - | 1 | 279 | 4309752 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPS28| Gene Symbol: RPS28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031.4 | NP_001022.1 | 1 | 300 | ||
| GenBank mRNA | AK301638.1 | - | 2 | 729 | ||
| BC021239.2 | - | 1 | 279 | 4309752 | ||