siRNA Details
| Gene Symbol: LOC101929601 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005251121.2 | XP_005251178.1 | 1 | 366 | ||
| Gene Symbol: LOC100288562 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002342862.5 | XP_002342903.1 | 1 | 366 | ||
| Gene Symbol: LOC101929627 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005251122.2 | XP_005251179.1 | 1 | 366 | ||
| Gene Symbol: LOC107986879 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017014126.1 | XP_016869615.1 | 1 | 366 | ||

LOC101929601
Genomic Map