siRNA Details
| Gene Symbol: PLXNC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005761.2 | NP_005752.1 | 10 | 2323 | ||
| XM_006719186.3 | XP_006719249.1 | 10 | 2350 | |||
| XM_011537730.2 | XP_011536032.1 | 10 | 2350 | |||
| XM_011537731.2 | XP_011536033.1 | 10 | 2350 | |||
| XM_017018671.1 | XP_016874160.1 | 10 | 2350 | |||
| XM_017018672.1 | XP_016874161.1 | 10 | 2350 | |||
| GenBank mRNA | AB208934.1 | - | 9 | 1004 | ||
| AF030339.1 | - | 10 | 2323 | |||

PLXNC1
Genomic Map