siRNA Details
| Gene Symbol: CCDC144B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036647.1 | 1 | 343 | |||
| GenBank mRNA | AK093811.1 | - | 1 | 343 | ||
| BC062767.1 | - | 1 | 337 | |||
| BC101585.1 | - | 1 | 258 | 8069091 | ||
| BC104815.1 | - | 1 | 258 | 8143818 | ||
| BC143561.1 | - | 1 | 258 | 9052072 | ||
| BX647826.1 | - | 1 | 334 | |||
| DB113689.1 | - | 1 | 343 | |||

CCDC144B
Genomic Map