siRNA Details
| Gene Symbol: CROCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014675.4 | NP_055490.4 | 20 | 2925 | ||
| XM_006711058.3 | XP_006711121.1 | 19 | 2716 | |||
| XM_011542468.1 | XP_011540770.1 | 20 | 2902 | |||
| XM_011542470.2 | XP_011540772.1 | 20 | 2902 | |||
| XM_017002913.1 | XP_016858402.1 | 20 | 2902 | |||
| XM_017002914.1 | XP_016858403.1 | 20 | 2902 | |||
| GenBank mRNA | AB007914.2 | - | 18 | 2564 | ||
| BC126911.1 | - | 17 | 2446 | |||
| BC126912.1 | - | 17 | 2446 | |||
| DQ139275.1 | - | 7 | 2282 | |||
| Gene Symbol: CROCCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK054737.1 | - | 1 | 1048 | ||
| AK090414.1 | - | 11 | 4684 | |||

CROCC
Genomic Map