siRNA Details
| Gene Symbol: ATP6AP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005765.2 | NP_005756.2 | 8 | 943 | ||
| GenBank mRNA | AF109363.1 | - | 8 | 905 | ||
| AF248966.1 | - | 8 | 948 | |||
| AF291814.1 | - | 8 | 930 | |||
| AK075382.1 | - | 8 | 1001 | |||
| AK293510.1 | - | 6 | 691 | |||
| AK296614.1 | - | 7 | 806 | |||
| AK297007.1 | - | 9 | 802 | |||
| AK297760.1 | - | 7 | 917 | |||
| AK314633.1 | - | 9 | 977 | |||
| AK315948.1 | - | 7 | 778 | |||
| AL049929.1 | - | 7 | 783 | |||
| AY038990.1 | - | 8 | 946 | |||
| AY429341.1 | - | 7 | 806 | |||
| BC010395.1 | - | 4 | 403 | |||
| BC084541.1 | - | 8 | 923 | 6575732 | ||
| Y17975.1 | - | 3 | 282 | |||

ATP6AP2
Genomic Map