siRNA Details
| Gene Symbol: RFC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017019779.1 | XP_016875268.1 | 9 | 997 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RFC5| Gene Symbol: RFC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017019779.1 | XP_016875268.1 | 9 | 997 | ||