siRNA Details
| Gene Symbol: RNF224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190228.1 | NP_001177157.1 | 3 | 608 | ||
| XM_011518914.2 | XP_011517216.1 | 2 | 498 | |||
| GenBank mRNA | BM921684.1 | - | 3 | 609 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF224| Gene Symbol: RNF224 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190228.1 | NP_001177157.1 | 3 | 608 | ||
| XM_011518914.2 | XP_011517216.1 | 2 | 498 | |||
| GenBank mRNA | BM921684.1 | - | 3 | 609 | ||