siRNA Details
| Gene Symbol: NOS2P3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC044655.1 | - | 4 | 2191 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NOS2P3| Gene Symbol: NOS2P3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC044655.1 | - | 4 | 2191 | ||