siRNA Details
| Gene Symbol: SLC35G5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054028.1 | NP_473369.1 | 1 | 1096 | ||
| GenBank mRNA | AJ291677.1 | - | 1 | 1096 | ||
| BC131696.1 | - | 1 | 1063 | 40120574 | ||
| Gene Symbol: SLC35G4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282300.1 | NP_001269229.1 | 1 | 1013 | ||

SLC35G5
Genomic Map