siRNA Details
| Gene Symbol: GOLGA8K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282493.1 | NP_001269422.1 | 12 | 1075 | ||
| Gene Symbol: GOLGA8H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282490.1 | NP_001269419.1 | 12 | 1073 | ||
| Gene Symbol: GOLGA8T | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033933.1 | 12 | 1095 | |||

GOLGA8K
Genomic Map