siRNA Details
Gene Symbol: LOC100996432 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136571.1 | 2 | 133 | |||
GenBank mRNA | DB073469.1 | - | 2 | 110 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100996432 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136571.1 | 2 | 133 | |||
GenBank mRNA | DB073469.1 | - | 2 | 110 |