siRNA Details
Gene Symbol: FAM230B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108107.1 | 9 | 1393 | |||
GenBank mRNA | AK128837.1 | - | 3 | 1206 |
Gene Symbol: LOC100996335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136559.1 | 8 | 1178 |
Gene Symbol: LOC100996432 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136571.1 | 8 | 1258 |
Gene Symbol: LOC105369189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030128.1 | XP_016885617.1 | 8 | 2054 |
