siRNA Details
| Gene Symbol: FAM153C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC101338.1 | - | 9 | 638 | 40025116 | |
| BC101341.2 | - | 9 | 638 | 40025123 | ||
| BC146883.1 | - | 9 | 850 | 9056811 | ||
| BC146888.1 | - | 9 | 850 | 9056816 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM153C| Gene Symbol: FAM153C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC101338.1 | - | 9 | 638 | 40025116 | |
| BC101341.2 | - | 9 | 638 | 40025123 | ||
| BC146883.1 | - | 9 | 850 | 9056811 | ||
| BC146888.1 | - | 9 | 850 | 9056816 | ||