siRNA Details
| Gene Symbol: LOC101929574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120661.1 | 2 | 540 | |||
| GenBank mRNA | BI439066.1 | - | 2 | 540 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929574| Gene Symbol: LOC101929574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120661.1 | 2 | 540 | |||
| GenBank mRNA | BI439066.1 | - | 2 | 540 | ||