siRNA Details
| Gene Symbol: LOC441155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271675.1 | NP_001258604.1 | 1, 2 | 231 | ||
| GenBank mRNA | AK302514.1 | - | 1, 2 | 188 | ||
| DB093153.1 | - | 1, 2 | 233 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC441155| Gene Symbol: LOC441155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271675.1 | NP_001258604.1 | 1, 2 | 231 | ||
| GenBank mRNA | AK302514.1 | - | 1, 2 | 188 | ||
| DB093153.1 | - | 1, 2 | 233 | |||