siRNA Details
| Gene Symbol: RBM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005777.2 | NP_005768.1 | 3 | 644 | ||
| XM_005264784.1 | XP_005264841.1 | 3 | 919 | |||
| XM_005264787.2 | XP_005264844.1 | 1 | 73 | |||
| XM_006712916.1 | XP_006712979.1 | 3 | 744 | |||
| XM_017005496.1 | XP_016860985.1 | 1 | 73 | |||
| XM_017005499.1 | XP_016860988.1 | 3 | 919 | |||
| XM_017005500.1 | XP_016860989.1 | 3 | 919 | |||
| XM_017005501.1 | XP_016860990.1 | 3 | 919 | |||
| GenBank mRNA | AF042857.1 | - | 3 | 497 | ||
| AF069517.1 | - | 3 | 600 | |||
| AF091264.1 | - | 3 | 529 | |||
| AK291719.1 | - | 3 | 644 | |||
| AK303371.1 | - | 3 | 537 | |||
| BX648535.1 | - | 3 | 821 | |||
| U50839.1 | - | 2 | 430 | |||

RBM6
Genomic Map