siRNA Details
| Gene Symbol: RBM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005778.3 | NP_005769.1 | 16 | 1476 | ||
| NR_036627.2 | 15 | 1402 | ||||
| XM_011533261.1 | XP_011531563.1 | 14 | 1254 | |||
| XM_011533262.1 | XP_011531564.1 | 16 | 1476 | |||
| XM_017005503.1 | XP_016860992.1 | 16 | 1458 | |||
| XM_017005505.1 | XP_016860994.1 | 15 | 1369 | |||
| XM_017005506.1 | XP_016860995.1 | 13 | 1219 | |||
| GenBank mRNA | AB208813.1 | - | 16 | 3962 | ||
| AF091263.1 | - | 16 | 1449 | |||
| AF103802.4 | - | 16 | 1427 | |||
| AK297249.1 | - | 15 | 1356 | |||
| AK297445.1 | - | 16 | 1331 | |||
| AK302766.1 | - | 15 | 1284 | |||
| AK314032.1 | - | 16 | 1431 | |||
| BC002957.1 | - | 16 | 1422 | |||
| KU178505.1 | - | 15 | 1301 | |||
| KU178506.1 | - | 14 | 1227 | |||
| U23946.1 | - | 16 | 1370 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_148930, AK146880, AJ309168, AK134183, BC031899, and others.

RBM5
Genomic Map