siRNA Details
| Gene Symbol: LHFPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005779.2 | NP_005770.1 | 5 | 1297 | ||
| XM_006714515.1 | XP_006714578.1 | 4 | 984 | |||
| GenBank mRNA | AK125227.1 | - | 2 | 794 | ||
| AY309920.1 | - | 2 | 696 | |||
| BC136335.1 | - | 2 | 648 | 9020322 | ||
| BC136336.1 | - | 2 | 648 | 9020323 | ||
| CR936660.1 | - | 3 | 934 | |||
| D86961.1 | - | 2 | 516 | |||

LHFPL2
Genomic Map