siRNA Details
| Gene Symbol: TSPY8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243721.1 | NP_001230650.1 | 4 | 714 | ||
| XM_005272744.4 | XP_005272801.1 | 4 | 728 | |||
| XM_011544005.2 | XP_011542307.1 | 4 | 721 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSPY8| Gene Symbol: TSPY8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243721.1 | NP_001230650.1 | 4 | 714 | ||
| XM_005272744.4 | XP_005272801.1 | 4 | 728 | |||
| XM_011544005.2 | XP_011542307.1 | 4 | 721 | |||