siRNA Details
Gene Symbol: ZNF710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005254906.3 | XP_005254963.1 | 1 | 84 | ||
XM_011521540.2 | XP_011519842.1 | 1 | 84 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005254906.3 | XP_005254963.1 | 1 | 84 | ||
XM_011521540.2 | XP_011519842.1 | 1 | 84 |