siRNA Details
| Gene Symbol: LINC01582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120325.1 | 2 | 113 | |||
| GenBank mRNA | DB041417.1 | - | 2 | 535 | ||
| HY026416.1 | - | 2 | 114 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01582| Gene Symbol: LINC01582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120325.1 | 2 | 113 | |||
| GenBank mRNA | DB041417.1 | - | 2 | 535 | ||
| HY026416.1 | - | 2 | 114 | |||