siRNA Details
| Gene Symbol: ALOX12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011523780.2 | XP_011522082.1 | 1 | 1876 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALOX12| Gene Symbol: ALOX12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011523780.2 | XP_011522082.1 | 1 | 1876 | ||