siRNA Details
| Gene Symbol: TSHZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308210.1 | NP_001295139.1 | 2 | 3400 | ||
| NM_005786.5 | NP_005777.3 | 2 | 3501 | |||
| XM_005266641.3 | XP_005266698.1 | 2 | 3771 | |||
| GenBank mRNA | AB210042.1 | - | 1 | 5596 | ||
| AF039698.1 | - | 1 | 2299 | |||
| AK127042.1 | - | 1 | 4242 | |||
| AK223545.1 | - | 1 | 2097 | |||
| AL833321.1 | - | 2 | 3069 | |||
| BC024152.2 | - | 1 | 1988 | |||

TSHZ1
Genomic Map