siRNA Details
| Gene Symbol: RPL13AP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003932.2 | 1 | 363 | |||
| Gene Symbol: RPL13A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270491.1 | NP_001257420.1 | 4, 5 | 341 | ||
| NM_012423.3 | NP_036555.1 | 5, 6 | 417 | |||
| NR_073024.1 | 5, 6 | 407 | ||||
| GenBank mRNA | AB082924.1 | - | 2, 3 | 196 | ||
| AK130605.1 | - | 5, 6 | 364 | |||
| AK222747.1 | - | 5, 6 | 343 | |||
| AK291120.1 | - | 5, 6 | 363 | |||
| BC000514.2 | - | 5, 6 | 346 | 2822802 | ||
| BC001675.2 | - | 1 | 344 | 2961667 | ||
| BC001836.2 | - | 1 | 344 | 2961667 | ||
| BC004900.2 | - | 4, 5 | 337 | |||
| BC013230.1 | - | 1 | 339 | |||
| BC032107.2 | - | 5, 6 | 352 | |||
| BC062537.1 | - | 4, 5 | 343 | 4908022 | ||
| BC065236.1 | - | 1 | 351 | 6161599 | ||
| BC070223.1 | - | 5, 6 | 363 | 6736442 | ||
| BC071929.1 | - | 4, 5 | 347 | 5808487 | ||
| BC105605.1 | - | 1 | 357 | 6726477 | ||
| BG536330.1 | - | 5, 6 | 364 | |||
| BQ069994.1 | - | 2 | 268 | |||
| Gene Symbol: RPL13AP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026712.1 | 1 | 363 | |||
 Back To Top
 Back To Top
 RPL13AP20
RPL13AP20 Genomic Map
Genomic Map Back To Top
 Back To Top