siRNA Details
| Gene Symbol: LOC728673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126047.1 | 2 | 2022 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC728673| Gene Symbol: LOC728673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126047.1 | 2 | 2022 | |||