siRNA Details
| Gene Symbol: GLIDR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126045.1 | 2, 3 | 554 | |||
| GenBank mRNA | BC000869.1 | - | 2, 3 | 408 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GLIDR| Gene Symbol: GLIDR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126045.1 | 2, 3 | 554 | |||
| GenBank mRNA | BC000869.1 | - | 2, 3 | 408 | ||