siRNA Details
| Gene Symbol: NBPF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278141.1 | NP_001265070.1 | 37 | 5998 | ||
| GenBank mRNA | AF131738.1 | - | 2 | 763 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NBPF12| Gene Symbol: NBPF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278141.1 | NP_001265070.1 | 37 | 5998 | ||
| GenBank mRNA | AF131738.1 | - | 2 | 763 | ||