siRNA Details
| Gene Symbol: CXorf36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176819.3 | NP_789789.2 | 5 | 1320 | ||
| XM_005272670.1 | XP_005272727.1 | 4 | 1121 | |||
| XM_006724559.1 | XP_006724622.1 | 4 | 1043 | |||
| GenBank mRNA | AK094877.1 | - | 3 | 545 | ||
| AK294472.1 | - | 4 | 889 | |||
| DA378381.1 | - | 2 | 414 | |||

CXorf36
Genomic Map