siRNA Details
| Gene Symbol: TRIM13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007278.2 | NP_001007279.1 | 4 | 960 | ||
| NM_005798.4 | NP_005789.2 | 3 | 761 | |||
| NM_052811.3 | NP_434698.1 | 3 | 846 | |||
| NM_213590.2 | NP_998755.1 | 2 | 1296 | |||
| GenBank mRNA | AF220127.1 | - | 3 | 701 | ||
| AF241850.1 | - | 3 | 719 | |||
| AJ224819.1 | - | 2 | 646 | |||
| AK314496.1 | - | 3 | 805 | |||
| AL360196.1 | - | 3 | 701 | |||
| AY191002.1 | - | 3 | 794 | |||
| AY455758.1 | - | 2 | 1271 | |||
| AY764035.1 | - | 4 | 930 | |||
| BC003579.1 | - | 3 | 807 | 3538675 | ||
| BC063407.1 | - | 2 | 1238 | 5212073 | ||

TRIM13
Genomic Map