siRNA Details
Gene Symbol: LOC401286 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_117091.1 | 3 | 2110 | |||
GenBank mRNA | AK123853.1 | - | 1 | 652 | ||
AK127120.1 | - | 3 | 2110 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC401286 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_117091.1 | 3 | 2110 | |||
GenBank mRNA | AK123853.1 | - | 1 | 652 | ||
AK127120.1 | - | 3 | 2110 |