siRNA Details
| Gene Symbol: AFDN-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027906.1 | 1 | 416 | |||
| GenBank mRNA | AB016898.1 | - | 1 | 329 | ||
| AK301419.1 | - | 1 | 408 | |||
| AK307980.1 | - | 2 | 70 | |||
| DC331926.1 | - | 1 | 416 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AFDN-AS1| Gene Symbol: AFDN-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027906.1 | 1 | 416 | |||
| GenBank mRNA | AB016898.1 | - | 1 | 329 | ||
| AK301419.1 | - | 1 | 408 | |||
| AK307980.1 | - | 2 | 70 | |||
| DC331926.1 | - | 1 | 416 | |||