siRNA Details
| Gene Symbol: DDX39A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005804.3 | NP_005795.2 | 7, 8 | 979 | ||
| XM_006722606.3 | XP_006722669.1 | 8, 9 | 1104 | |||
| XM_011527620.1 | XP_011525922.1 | 7, 8 | 998 | |||
| XM_011527621.2 | XP_011525923.1 | 7, 8 | 923 | |||
| GenBank mRNA | AK301847.1 | - | 8, 9 | 1062 | ||
| AL512698.1 | - | 1, 2 | 1185 | |||
| BC001009.2 | - | 7, 8 | 916 | 2820926 | ||
| BM792110.1 | - | 1, 2 | 112 | |||
| U90426.1 | - | 7, 8 | 956 | |||

DDX39A
Genomic Map