siRNA Details
| Gene Symbol: PRG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127708.2 | NP_001121180.2 | 6 | 2811 | ||
| NM_001127709.2 | NP_001121181.2 | 5 | 2655 | |||
| NM_001127710.2 | NP_001121182.2 | 4 | 2532 | |||
| NM_001303232.1 | NP_001290161.1 | 6 | 2805 | |||
| NM_005807.4 | NP_005798.3 | 7 | 2934 | |||
| XM_017000002.1 | XP_016855491.1 | 4 | 2951 | |||
| XM_017000003.1 | XP_016855492.1 | 3 | 2518 | |||
| GenBank mRNA | AK057197.1 | - | 5 | 770 | ||
| AK131434.1 | - | 6 | 1505 | |||
| AK301343.1 | - | 8 | 1967 | |||
| U70136.1 | - | 6 | 2916 | |||

PRG4
Genomic Map