siRNA Details
| Gene Symbol: GDF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005811.3 | NP_005802.1 | 3 | 1099 | ||
| XM_006719194.3 | XP_006719257.1 | 3 | 1101 | |||
| GenBank mRNA | AF028333.1 | - | 3 | 980 | ||
| AF100907.1 | - | 3 | 1100 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GDF11| Gene Symbol: GDF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005811.3 | NP_005802.1 | 3 | 1099 | ||
| XM_006719194.3 | XP_006719257.1 | 3 | 1101 | |||
| GenBank mRNA | AF028333.1 | - | 3 | 980 | ||
| AF100907.1 | - | 3 | 1100 | |||