siRNA Details
Gene Symbol: C16orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522462.2 | XP_011520764.1 | 1 | 119 | ||
GenBank mRNA | BI831003.1 | - | 1 | 123 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C16orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522462.2 | XP_011520764.1 | 1 | 119 | ||
GenBank mRNA | BI831003.1 | - | 1 | 123 |